Learn More
Masterflex™ Stainless Steel Barbed Fitting NPT Male Pipe Adapter
Click to view available options
Dimensioni filo:
1.5 in. NPT(M)
1/2 in. NPT(M)
1/4 in. NPT(M)
3/8 in. NPT(M)
Dimensioni dei tubi:
1 1/2 in. I.D.
1/16 in. I.D.
1/2 in. I.D.
1/4 in. I.D
1/4 in. I.D.
3/8 in. I.D.
Dimensione della confezione:
Pezzo
Descrizione
- Available in a variety of materials and configurations to meet your need
- Adapts from a NPT male thread to a hose barb
Specifica
Specifica
| Materiale | Stainless Steel |
| Overall Length (English) | 1 27/32 in. |
| Dimensioni dei tubi | 1/4 in. I.D |
| Flessibile flangiato | 1/4 in. |
| Porta di uscita | 1/2 in. NPT(M) |
| Dimensione della porta | 1/4 in. |
| Dimensioni filo | 1/2 in. NPT(M) |
| Larghezza (sistema imperiale) | 7/8 in. |
| Product Type | Barbed Fitting NPT Male Pipe Adapter |