Learn More
IKA™ Stainless Steel Beater
Sfoglia Tutto IKA Prodotti
Click to view available options
:
For Use With: Pulverizing Substances with a °Mohs Hardness up to 9
Specifica
Specifica
| Product Type | Beater |
| Da utilizzare con (applicazione) | For Pulverizing Substances with a Mohs Hardness up to 9 |
| Durezza | Up to 9 °Mols |
| Materiale | Stainless Steel |