Learn More
Masterflex™ B/T™ Analog Air-Powered Drive
Click to view available options
Portata:
11.1 gal./min.
Dimensione della confezione:
Pezzo
Descrizione
- Flow rate 0.71/2.1 to 42 LPM with B/T™ tubing—flow rate depends on drive rpm and tubing size
- Rapid-Load™ pump head occlusion mechanism slides occlusion bed out of the way for easy tube loading and improved tubing occlusion
- Accepts B/T™ PerfectPosition™ pump tubing sizes B/T™ 87 and B/T™ 91
- Interlock switch shuts off power to stop drive when door is opened
Specifica
Specifica
| Certificazione/Conformità | CE II 3 G c IIC T6, EN 13463-1 & EN 13463-5 (EU ATEX Directive), EN 809 (EU Machinery Directive) |
| Control Type | Variable Speed |
| Tipo di display | Digital |
| Valori nominali armadio | IP56 |
| Portata | 11.1 gal./min. |
| Maximum Pressure | 35 (B/T 87), 30 (B/T 91) psi |
| Motor Speed | 321 rpm |
| Finitura | Epoxy-Powder |
| Da utilizzare con | Air-powered drive in locations where electricity is unsafe or impractical |
| Da utilizzare con (applicazione) | Bulk-transfer |
| Vedi altri risultati |