Learn More
Nickel Electro™ Stainless Steel Flask Clip
Codice prodotto. 11479629 Sfoglia Tutto Nickel-Electro Prodotti
Cambia vista
Click to view available options
Capacità:
1 L Conical Flask
100 mL Conical Flask
2 L Conical Flask
2.8 L Fernbach flask
25 mL Conical Flask
250 mL Conical Flask
50 mL Conical Flask
500 mL Conical Flask
Dimensione della confezione:
Pezzo
Codice prodotto. Capacità unitSize
11479629 1 L Conical Flask Pezzo
11776577 2 L Conical Flask Pezzo
11736577 25 mL Conical Flask Pezzo
11766577 250 mL Conical Flask Pezzo
11469629 500 mL Conical Flask Pezzo
11746577 50 mL Conical Flask Pezzo
11756577 100 mL Conical Flask Pezzo
12778968 2.8 L Fernbach flask Pezzo
8 options
Codice prodotto. 11479629
Marca: Nickel Electro™ FC1000
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
For personalised clip arrangement, clips are supplied with all necessary fixings to enable them to be attached to the Perforated Tray. Nickel Electro™ Stainless Steel Flask Clip is suitable for flasks and can be autoclaved.
  • Highly flexible elastic design allowing glassware to easily slide into the clip and yet remain secure during mixing
  • No sharp edges to cut or score glassware
  • Can be autoclaved
  • Plastic base
  • Easy to assemble and remove Flask Clips from Perforated tray
Specifica
Capacità 1 L Conical Flask
Product Type Flask Clip
Materiale Stainless Steel