Learn More
Gilson™ Stainless Steel Connectors
Sfoglia Tutto Gilson Prodotti
Click to view available options
Diametro (sistema metrico):
0.6 mm
0.8 mm
1.1 mm
Dimensione della confezione:
pezzi 5
Descrizione
- Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
- PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Specifica
Specifica
| Diametro (sistema metrico) | 0.8 mm |
| Materiale | Stainless Steel |
| Quantità confezione | 5 Set |
| Include | Metal sleeve |
| Product Type | Connector |