Learn More
AMSTA Rostfritt Stainless Steel Container with Lid
Codice prodotto. 11916423
Click to view available options
Capacità (sistema metrico):
8 L
20 L
25 L
30 L
50 L
100 L
150 L
Include:
Lid
Lid and 3 Clamps
Lid and 4 Clamps
Lid and 6 Clamps
Lid and Tap
Lid, Tap, and 3 Clamps
Lid, Tap, and 4 Clamps
Lid, Tap, and 6 Clamps
Dimensione della confezione:
Pezzo
Codice prodotto. 11916423
Marca: AMSTA Rostfritt 50608201+5051310X3+50806
High-quality stainless steel container, available in various sizes with or without clamps and tap.
  • Quality
  • Invisible weld seams
  • Pharmaceutical grade finish
  • Sustainable production
  • Made in Sweden
Specifica
Product Type Can
Capacità (sistema metrico) 20 L
Colore Silver
Materiale Stainless Steel
Finitura Ra 0.2 to 0.3 μm
Include Lid and 3 Clamps
Altezza (sistema metrico) 397 mm
Diametro (sistema metrico) esterno 270 mm
Peso (sistema metrico) 4.2 kg