Learn More
Amsta Rostfritt Stainless Steel Containers
Click to view available options
Volume (sistema metrico):
1 L
2 L
3 L
5 L
6 L
10 L
50 L
Include:
3 clamps, without lid
With Lid
Without Lid
Dimensione della confezione:
Pezzo
Specifica
Specifica
| Product Type | Container with Lid |
| Materiale | Stainless Steel |
| Dimensioni (L x P) | 220 x 300 mm |
| Include | 3 clamps, without lid |
| Volume (sistema metrico) | 10 L |