Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class M1 with certificate
Codice prodotto. 15535139 Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Peso (sistema metrico):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
Dimensione della confezione:
1 unità
Codice prodotto. 15535139
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifica
Classificazione M1
Peso (sistema metrico) 1 g to 5 kg
Massa 1 g to 5 kg