Learn More
Fisherbrand™ Stainless Steel Round Bottom Weighing Scoops
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Lunghezza (sistema metrico):
70 mm
90 mm
105 mm
120 mm
Dimensione della confezione:
pezzi 5
Descrizione
- Well-suited for pharmaceutical and environmental sampling
- Ideal for weighing and handling of samples
- Great for weighing messy ingredients, tiny objects and portions
- Great accessory to have if you own a weighing scale
- Perfect for weighing anything from liquid to gems and other materials
Specifica
Specifica
| Forma | Cylindrical |
| Product Type | Scoop |
| Larghezza (sistema metrico) | 30 mm |
| Dimensioni (L x P) | 70 x 30 mm |
| Materiale | Stainless Steel |
| Lunghezza (sistema metrico) | 70 mm |