Learn More
Aalborg™ Griglia in acciaio inox
Codice prodotto. 11802340
Click to view available options
Da utilizzare con (apparecchiatura):
Flussometro gas/liquido 65 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 11802340
Marca: Aalborg™ P16S2VA0
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Aalborg™ Stainless Steel Support Frame for 65mm P-meter is for use with single tube flow meters, which are available with built in valves.
  • Rib-guided or fluted metering tubes facilitate stable and accurate readings
  • Magnifier lens in front shield to enhance reading resolution
  • Interchangeability of Flow Tubes and floats
  • Ease of installation and exchange of flow tubes
  • Glass flow tubes are prevented from turning during the tightening phase of the assembly procedure
  • Chemical compatibility
  • Simple means of panel mounting
  • Frame for P-meter
Specifica
Da utilizzare con (apparecchiatura) Flussometro gas/liquido 65 mm
Tipo Telaio
Larghezza (sistema metrico) 65 mm