Learn More
Nickel Electro™ Coperchio a tetto in acciaio inox
Codice prodotto. 11439519 Sfoglia Tutto Nickel-Electro Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
2 and 4 L Unstirred Baths
5 and 14 L Unstirred Baths
8 and 10 L Unstirred/Shaking Baths
Bagnomaria con agitazione e senza agitazione da 22 e 28 l
Dimensione della confezione:
Pezzo
Codice prodotto. 11439519
Marca: Nickel Electro™ SL18
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Obtain optimum results keeping samples in a water bath free from contamination and saving energy. Designed allowing for full use of whole width of a Clifton™ water bath, Clifton™ Stainless Steel Gable Lids help reduce energy consumption and running costs.
  • Designed allowing full use of whole width of a Clifton™ water bath
  • Helps reduce evaporation
  • Asists in keeping samples in a water bath free from contamination and saves energy
  • Allows extended use of the bath, right into the edges of the tank
  • Lid design ensures all condensate returns back into the bath
  • Insulated handles for safe use
  • Reduces energy consumption and running costs for your Clifton water bath
  • Lowest height of the gable being 45mm above the bath
Specifica
Da utilizzare con (apparecchiatura) 8 and 10 L Unstirred/Shaking Baths
Materiale Stainless Steel