Learn More
OHAUS™ Stainless Steel Lab Frame
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Size:
305 x 1829 x 1219 mm (LxWxH)
457 x 1219 x 1219 mm (LxWxH)
457 x 1219 x 610 mm (LxWxH)
457 x 610 x 610 mm (LxWxH)
551 x 1829 x 1905 mm (LxWxH)
Dimensione della confezione:
Pezzo
Descrizione
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Dependable connectors secure rods and clamps to meet your requirements and five unique holders in a variety of sizes and materials accommodate different rods.
Specifica
Specifica
| Tipo | Lab Frame Kit |
| Materiale | Stainless Steel |
| Descrizione | Clamp, Lab Frame, Steel, CLR-FRAMESH |
| Altezza (sistema metrico) | 1,905.000 mm |
| Altezza (sistema imperiale) | 75.00 in. |
| Lunghezza (sistema imperiale) | 21.70 in. |
| Lunghezza (sistema metrico) | 551.00 mm |
| Size | 551 x 1829 x 1905 mm (LxWxH) |
| Larghezza (sistema imperiale) | 72.00 in. |
| Larghezza (sistema metrico) | 1,829.00 mm |