Learn More
Grant Instruments™ Stainless Steel Lid
Click to view available options
Da utilizzare con (apparecchiatura):
ST12 tank
ST18, ST26 tanks
ST38 tank
Dimensione della confezione:
Pezzo
Descrizione
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
- Stainless steel construction
- Gabled with plastic handle
- For use with Grant Stirred Water Baths and Heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
- For continuous use with water above 90°C.
Specifica
Specifica
| Descrizione | Length: 375 |
| Da utilizzare con (apparecchiatura) | ST18, ST26 tanks |
| Gamma di temperatura | 90°C |
| Materiale | Stainless Steel |