Learn More
Aalborg™ Stainless Steel N2 Mass Flowmeter
Codice prodotto. 11757889
Click to view available options
Tipo:
Mass Flowmeter
Unit Size:
Pezzo
Regulatory: This product is under eco-contribution tax.
Product Code. 11757889
Brand: Aalborg™ GFC17AVAL6C0/3NC09SS
Only null left
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
Specifications
Tipo Mass Flowmeter
Accuracy ±5%
Certificazione/Conformità NIST calibration certificate
Intervallo di portata 0 to 5 LPM
Dettagli alimentazione 12 V to 200 mA max
Precisione ±1.5 %
Bilancia Full scale
Temperatura (sistema metrico) 5°C to 50°C