Learn More
Azpack™ Clip per tubi in acciaio inox
Click to view available options
Diametro (sistema metrico):
10 to 14 mm
13 to 17 mm
18 to 22 mm
Dimensione della confezione:
pezzi 5
Descrizione
- Stainless steel
- Double-banded ring for even pressure
- Nut and bolt included to apply proper compression
Specifica
Specifica
| Tipo | Tubing Clamp |
| Materiale | Stainless Steel (H10 304, A2) |
| Include | Tightening bolt |
| Diametro (sistema metrico) | 13 to 17 mm |
| Jaw Type | Double-banded Ring |