Learn More
Cole-Parmer™ Stainless Steel Union Elbow
Click to view available options
Materiale:
Stainless Steel (316)
Dimensione della confezione:
Pezzo
Descrizione
- Very good chemical resistance.
- Rates to 220psi at 77°F (25°C).
- Usable from 32 to 212°F (0 to +100°C).
- Compression fittings work well with rigid tubing such as PTFE or polypropylene.
- A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
- Compression connector family is ideal when working with rigid tubing.
- When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Specifica
Specifica
| Descrizione | Compression connection |
| Materiale | Stainless Steel (316) |
| Metodo di sterilizzazione | Autoclave, Ethylene Oxide or Gamma |
| Product Type | Union Elbow |