Learn More
Testo™ Stainless Steel Waterproof Food Probe
Marca: Testo™ 0602 2292
Dettagli aggiuntivi : Peso : 0.11100kg
Specifica
Food Probe, 4 dia. x 125mm L Probe Shaft, 3.2 dia. x 50mm L Probe Shaft Tip, Stainless Steel, –60 to 400°C Measuring Range, 7 sec. Response Time, Waterproof | |
EN 60584-2 | |
Stainless Steel | |
7 sec. |