Learn More
Fisherbrand™ Standardization Pipet Tips
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Sterilità:
Sterile
Colore:
Natural
Yellow
Imballaggio:
Stack Rack, 5 x 200
Dimensione della confezione:
pezzi 1000
Descrizione
Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip
- Soft pliable tip collar allows this tip to fit on pipets made by various manufacturers
- Lower cost and increased convenience by purchasing fewer products
- Available in a variety of packaging options
- Eliminate the need to buy pipetter-specific tips
- Certified by independent laboratory
- Nonsterile: RNase, DNase, DNA and pyrogen free
Packaging Code -- BP: Bulk Pack; RI: Rack Inserts; HR: Hinged Rack; HRS: Hinged Rack; HR/T: Hinged Rack, Trace Metal-free; LR: Lift-off Rack; LRS: Lift-off Rack; SR: Stack Rack; IWS: Individually Wrapped.
Specifica
Specifica
| Volume (sistema metrico) | 0.1 to 200 μL |
| Sterilità | Sterile |
| Colore | Yellow |
| Autoclavabile | Autoclavable |
| Lunghezza (sistema metrico) | 5cm |
| Dichiarazioni di pulizia | DNase, RNase, DNA, PCR inhibitor, and Pyrogen Free |
| Formato | In rack |
| Stile punta | Universal |
| Imballaggio | Stack Rack, 5 x 200 |
| Materiale | Polipropilene |