Learn More
Stathmin 1 Antibody (STMN1/8001R), R&D Systems™
Product Code. 18606914 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18606914 20 μg 20µg
18652914 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18606914 Supplier Novus Biologicals Supplier No. NBP32034620ug
 Restricted Item
Spedizione stimata: 27-11-2026
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
Stathmin 1 Monoclonal specifically detects Stathmin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Antigen Stathmin 1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone STMN1/8001R
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias C1orf215, chromosome 1 open reading frame 215, Lag, Leukemia-associated phosphoprotein p18, Metablastin, MGC138869, Oncoprotein 18, Op18, OP18MGC138870, phosphoprotein 19, Phosphoprotein p19, pp17, PP19, PR22, prosolin, Protein Pr22, SMNLAP18FLJ32206, stathmin, stathmin 1, stathmin 1/oncoprotein 18, transmembrane protein C1orf215
Host Species Rabbit
Immunogen Recombinant fragment of human Stathmin 1 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cognition and Behavior, Immunology, Microglia Markers, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3925
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Product Type Antibody
Form Purified
Isotype IgG κ
Product Line Novus