Learn More
Abeba™ Static Control 31392 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20345:2011 S2 SRA
- Microfiber
- White
- Composite toe cap
- PU midsole
- Slip resistance special rubber outsole sole
- Replaceable acc Wave insole (article no. 3553)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with elastic band
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20345:2011 S2 SRA |
| Colore | White |
| Materiale | Microfiber |
| Size | 44 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Rubber |
| Stile | 31392 |
| Materiale dello zoccolo | Composite |