Learn More
Merck Stericup™ Quick Release Bottle Top Receiver Flasks
Codice prodotto. p-7156823 Sfoglia Tutto Merck Prodotti
Click to view available options
Volume (sistema metrico):
250 mL
500 mL
1000 mL
Dimensione della confezione:
pezzi 12
Codice prodotto. 15967996
Marca: Merck S200B02RE
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Stericup™ Quick Release Receiver Flask for Bottle Top Filtration includes a click-seal confidence cap that enables a secure grip in wet and dry conditions.
Steritop™ bottle top filter units provide a large filter area for rapid sterile filtration of aqueous fluids into narrow-mouth (33 mm) or wide-mouth (45 mm) bottles.
A small tab inside the funnel secures an optional prefilter in-place during filtration.
For added protection against contamination, the threads for bottle attachment have been slightly recessed in case the funnel accidentally touches a work surface.
TRUSTED_SUSTAINABILITY
Specifica
Raccordo uscita 45 mm
Volume (sistema metrico) 250 mL