Learn More
Bel-Art™ Sterileware™ Cross-Sectional Liquid Samplers
Sfoglia Tutto SP Scienceware Prodotti
Click to view available options
Capacità (sistema metrico):
80 mL
225 mL
Dimensione della confezione:
pezzi 20
Descrizione
- Provides effective cross-sectional sampling of liquids and light creams
- Reaches bottom of container while keeping hand clear of container top
- Diameter allows for use through drum bunghole or other small openings
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) samplers heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifica
Specifica
| Product Type | Liquid Sampler |
| Materiale | HDPE |
| Autoclavabile | Not Autoclavable |
| Capacità (sistema imperiale) | 7.5 oz. |
| Capacità (sistema metrico) | 225 mL |
| Quantità | 20/Pk. |
| Diametro (sistema metrico) esterno | 2.1 cm |
| Lunghezza (sistema metrico) | 110 cm |