Learn More
Bel-Art™ Sterileware™ Extra-Long, Bent Handle Spoons
Sfoglia Tutto SP Scienceware Prodotti
Click to view available options
Capacità (sistema metrico):
10 mL
20 mL
Dimensione della confezione:
pezzi 100
Descrizione
- Long handle allows sampling from a distance reducing the risk of contamination
- Handle is bent at bowl to facilitate easier retrieval of samples, including from within tall containers
- Made from FDA and EU conforming materials; safe for food, drug and cosmetic use
- High density polyethylene (HDPE) spoons heat sealed in individual polyethylene bags
- Gamma irradiated sterile (SAL 10^-6)
- Lot stamped for accurate tracing; shipped with a Certificate of Processing for Sterilization
Specifica
Specifica
| Quantità | 100/Pk. |
| Autoclavabile | Not Autoclavable |
| Capacità (sistema metrico) | 20 mL |
| Product Type | Spoon |
| Lunghezza (sistema metrico) maniglia | 30 cm |
| Materiale | HDPE |