Learn More
Buerkle™ SteriPlast™/LaboPlast™ Palette Knife Spatula
Sfoglia Tutto Bürkle Prodotti
Click to view available options
Sterilità:
Non-sterile
Sterile
Dimensione della confezione:
pezzi 100
Descrizione
- The dimensionally stable long edge of the plastic scraper is ideal for cutting viscous media.
- A small tip on the long edge can be used for ripping, for example packaging film.
- The scraper is also suitable for stirring and blending.
- PS, white
- Available either in a large package with 100 pieces or 100 pieces packaged individually and radiation sterilised
- For single use
- Clean room manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
Specifica
Specifica
| Quantità | 100 |
| Sterilità | Sterile |
| Autoclavabile | Not Autoclavable |
| Certificazione/Conformità | Complies with EU food rrequirements and FDA regulations |
| Monouso | Single-use |
| Product Type | Sampler |
| Lunghezza (sistema metrico) lama | 70 mm |
| Materiale | Polystyrene |
| Lunghezza (sistema metrico) | 192 mm |