Learn More
Sartorius Sterisart™ Gen 4 - Isolator Installation Kit
Sfoglia Tutto Sartorius Prodotti
Click to view available options
Tipo:
Isolator Installation Kit
Dimensione della confezione:
1 confezione
Descrizione
Sterisart™ Universal ∣ Gen 4 - Isolator Installation Kit
Contains:
- 1x stainless steel adapter
- 2x brass bolts to secure the housing
- 2x countersink screws M5 x 12
- 2x silicone o-rings (270 mm x 3 mm)
- 3x cylinder head screws M6 x 12
- 3x cylinder head screws M6 x 8
- 3x body washers (ø 6,4 x ø 18 x 1,6)
- 1x sensorboard protection cover
- 1x cylinder head screws M3 x 12
Specifica
Specifica
| Tipo | Isolator Installation Kit |