Learn More
Sampling Systems™ SteriWare™ Capsule Counting Trays
Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Sterilità:
Non-sterile
Sterile
Dimensione della confezione:
pezzi 20
Descrizione
- Our capsule counting trays makes accurate sorting and counting easy.
- They are made from robust, shatterproof PS.
- They are suitable for use with all capsules from Size 2 to Size 00.
- Use straight from the heat sealed PE bag.
- They are manufactured and packed under cleanroom conditions.
- Made from white food grade PS.
- Conforms to the latest FDA and EU requirements.
- BSE/TSE free.
- Either sterile or non sterile.
Specifica
Specifica
| Altezza (sistema metrico) | 12 mm |
| Lunghezza (sistema metrico) | 260 mm |
| Materiale | Polypropylene |
| Larghezza (sistema metrico) | 130 mm |
| Sterilità | Non-sterile |
| Tipo di imballaggio | Bag |