Learn More
Sampling Systems™ SteriWare™ Disposable HDPE Powder Lances
Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Capacità (sistema metrico):
75 mL
Dimensione della confezione:
pezzi 20
Descrizione
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Suitable for use in ATEX areas
- Push the sampler into the product, rotate the sampler then withdraw
- Deposit the sample into a suitable container
- Place the used sampler into its original plastic bag and dispose
- No cross contaminatin
- No cleaning required
Specifica
Specifica
| Materiale | HDPE |
| Include | 20 per box |
| Capacità (sistema metrico) | 75 mL |
| Sterilità | Sterile |
| Lunghezza (sistema metrico) | 400 mm |