Learn More
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief
BIOPHARMA
Codice prodotto. 11915636 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Capacità (sistema metrico):
1 mL
5 mL
10 mL
20 mL
Lunghezza (sistema metrico):
500 mm
1000 mm
Dimensione della confezione:
pezzi 20
Codice prodotto. 11915636
Marca: Sampling Systems™ 8060H1005
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Sampling Systems™ SteriWare™ HDPE Disposable Powder Thief is ideal for taking representative samples from known locations within the bulk powder.
  • Assembled and packed in a cleanroom
  • Individually Bagged
  • Made from FDA or EU conforming HDPE
  • BSE/TSE Free
  • Full Batch Traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • No cleaning is required before or after sampling
  • No cleaning validation (this is very expensive to set up and run)
  • Manufactured in a class 100,000 cleanroom environment (check individual sampler descriptions)
  • Peace of mind
  • Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)
  • Suitable for use in ATEX Zones 0, 1 and 2
  • Financial
  • Save time and money (it will cost more to clean a reusable sampler than to buy a disposable sampler)
Specifica
Materiale HDPE
Capacità (sistema metrico) 5 mL
Sterilità Non-sterile
Diametro (sistema metrico) 21 mm
Lunghezza (sistema metrico) 1000 mm