Learn More
Sampling Systems™ SteriWare™ LiquiDip Duo
BIOPHARMA
Codice prodotto. 15809581 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Capacità (sistema metrico):
80 mL
225 mL
Lunghezza (sistema metrico):
450 mm
1100 mm
Sterilità:
Non-sterile
Sterile
Dimensione della confezione:
pezzi 20
Codice prodotto. 15809581
Marca: Sampling Systems™ 8921H450
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Sampling Systems™ SteriWare™ LiquiDip Duo is ideal for taking cross sectional or point samples of liquids.
  • Assembled and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
  • BSE/TSE free
  • Suitable for taking cross sectional and point samples
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • Available in 2 different lengths
  • Push the sampler into the product, ensuring the top and bottom are in the open position
  • Push the sampler against the base of the container to close the valve
  • Remove the sampler and pour out the liquid into a suitable container
  • No cross contamination
  • No cleaning required
Specifica
Product Type Liquid Sampler
Materiale HDPE
Include Valve at both ends
Capacità (sistema metrico) 80 mL
Sterilità Non-sterile
Diametro (sistema metrico) esterno 21 mm
Lunghezza (sistema metrico) 450 mm