Learn More
Sampling Systems™ SteriWare™ PP White Disposable PharmaScoop™
Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Capacità (sistema metrico):
60 mL
500 mL
1000 mL
Sterilità:
Non-sterile
Sterile
Dimensione della confezione:
pezzi 100
pezzi 25
pezzi 50
Descrizione
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged in ultra-strong packing with easy to open design
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Large variety of different sizes available
- Good Manufacturing Practice (GMP) to keep your hands out of the product as much as possible
- Positioned the ergonomic handle high up on the scoop
- No Cross Contamination
- No Cleaning required
Specifica
Specifica
| Sterilità | Non-sterile |
| Capacità (sistema metrico) | 500 mL |
| Colore | White |
| Monouso | Single-use |
| Product Type | Disposable Scoop |
| Include | Individually Bagged |
| Materiale | Polypropylene |