Learn More
Sampling Systems™ SteriWare™ Sack and Bag Samplers
Shop All Sampling Systems Products
missing translation for 'orderingAttributeHoverText'
Diametro (sistema metrico) parte inferiore esterna:
25 mm
45 mm
Sterilità:
Non-sterile
Sterile
missing translation for 'unitSize'
pezzi 100
Description
- The SteriWare™ Sack and Bag Sampler is perfect for taking samples from sacks and bags.
- This sampler is ideal for fast quality checks through the sides of the sacks.
- The SteriWare™ Sack Sampler is available in two diameters, the narrow one is ideal for free flowing powders and for small products like pepper corns, the large diameter sampler is more suited to sampling nuts and beans.
- The SteriWare™ Sack Sampler has an ergonomic handle and specially designed blade.
- During sampling, the operator needs to hold a bag or bottle over the rear end of the sampler to catch the powder or granules as they slide through the sampler.
- Made from high quality HDPE which is food and FDA compliant grade.
- BSE/TSE free.
- Full batch documentation and certification supplied.
- Individually bagged and sold in economic boxes of 100.
Specifications
Specifications
| Materiale | HDPE |
| Sterilità | Sterile |
| Tipo di imballaggio | Box |
| Quantità | 100 Pk. |
| Diametro (sistema metrico) parte inferiore esterna | 25 mm |
| Lunghezza (sistema metrico) | 325 mm |