Learn More
Fisherbrand™ BS 2775 Natural Rubber Stopper With Inlet/Outlet Holes
Codice prodotto. 11527652 Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Materiale:
Rubber
Dimensione della confezione:
pezzi 48
Codice prodotto. 11527652
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Chemically resistant, flexible, tightly fitting natural rubber stopper with predrilled inlet/outlet holes. Fisherbrand™ BS 2775 Natural Rubber Stopper With Inlet/Outlet Holes work well for ventilated and vacuum glassware.
  • Tapered plug with predrilled inlet/outlet holes.
  • Fits a variety of test tubes, flasks and other laboratory glassware.
  • Chemically resistant.
  • Tight sealing.
Specifica
Da utilizzare con (applicazione) For laboratory use in bottles and flasks
Quantità 48/Pk.
Materiale Rubber
Product Type Tapered Stopper (2 holes)