Learn More
Merck Strat-M™ Membrane
Sfoglia Tutto Merck Prodotti
Click to view available options
Colore:
White
Dimensione della confezione:
Pezzo
Descrizione
- Active pharmaceutical ingredients (API)
- Cosmetic actives
- Formulations
- Personal care products
- Pesticides and chemicals
- Predictive of diffusion in human skin for a wide range of compounds and formulations
- Exhibits differential permeability in the presence of enhancers and is compatible with common formulation formats, so it is appropriate for use during formulation optimization
Specifica
Specifica
| Colore | White |
| Materiale | Polyethersulfone, Polyolefin, Synthetic Lipid Post-treatment |
| Codice filtro | SKBM |
| Tipo di superficie | Plain |
| Spessore (sistema metrico) | 300 μm |
| Tipo | Transdermal Diffusion Test Model |
| Diametro (sistema metrico) | 2.5 cm |