Learn More
IBA GmbH™ Strep-Tactin™ Chromeo 546 Conjugate
Sfoglia Tutto IBA Lifesciences Prodotti
Click to view available options
:
Quantity: 50 μg
Descrizione
- For the detection of Strep-tag™ II and Twin-Strep-tag™ in Immunofluorescence, FACS and more
- Contains Strep-Tactin™ conjugated to Chromeo 546
- 1 x 50μg fluorescent conjugate
Specifica
Specifica
| Quantità | 50 μg |