Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Sfoglia Tutto IBA Lifesciences Prodotti
Click to view available options
Quantità:
1 mL
5 mL
5 x 0.2 mL
Dimensione della confezione:
1mL
pezzi 5
Descrizione
- Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
- To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifica
Specifica
| Quantità | 5 mL |
| Tipo di colonna | Gravity flow |
Esclusivamente per scopi di ricerca.