Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ resin
Codice prodotto. 15523077 Sfoglia Tutto IBA Lifesciences Prodotti
Click to view available options
Capacità (sistema metrico):
4 mL
20 mL
50 mL
1000 mL
Dimensione della confezione:
1000mL
20mL
4mL
50mL
Codice prodotto. 15523077
Marca: IBA Lifesciences 24010025
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Strep-Tactin™XT Superflow™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT Superflow™ is a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
  • In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
  • The suspension is suitable for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.
Specifica
Capacità (sistema metrico) 50 mL
Tipo Resin
Esclusivamente per scopi di ricerca.