missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Strontium ranelate
Click to view available options
Quantità:
5 g
25 g
100 g
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Materiale o nome chimico | Strontium ranelate |
Quantità | 25 g |
Peso formulazione | 513.49 |
InChI Key | KVJIIORUFXGTFN-UHFFFAOYSA-L |
SMILES | [Sr++].OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O.OC(=O)CN(CC(O)=O)C1=C(C#N)C(CC([O-])=O)=C(S1)C(O)=O |
IUPAC Name | strontium(2+) bis(2-{5-[bis(carboxymethyl)amino]-2-carboxy-4-cyanothiophen-3-yl}acetate) |
Molecular Weight (g/mol) | 770.16 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'