Learn More
Cole-Parmer™ Dry Block Heater Cover
Click to view available options
Size:
2 Biock System
3 Block or Dual Control
Dimensione della confezione:
Pezzo
Descrizione
- The heater covers ensure temperatures are maintained in your dry block heaters
- Model 36610-96 cover is suitable for use with two block heaters
- Model 36610-97 is suitable for use with three block or dual control heaters
- The extraction tool, 36610-98, makes removing blocks from the heater quick and effortless
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | 2-block System |
| Tipo | Safety Cover |
| Size | 2 Biock System |
Titolo del prodotto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido