Learn More
Cole-Parmer™ Drum for DR-200 Series Rotating Shaker System Drives
Click to view available options
N. di posizioni:
1
2
4
240
Da utilizzare con (apparecchiatura):
1.5 mL Microcentrifuge Tubes
250 mL Flasks or Bottles
Bottles
Clip Plates
One Bottle, Figure-8 Motion
Dimensione della confezione:
Pezzo
Descrizione
- Change your rotator's capacity with interchangeable drums
- Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifica
Specifica
| Product Type | Drum |
| Da utilizzare con (applicazione) | For changing your rotator's capacity |
| Da utilizzare con (apparecchiatura) | One Bottle, Figure-8 Motion |
| N. di posizioni | 1 |