Learn More
Cole-Parmer™ Stuart™ Heater Block Insert
Click to view available options
Dimensioni provetta:
Medium
Large
Dimensione della confezione:
Pezzo
Descrizione
- Safest way to heat round-bottom flasks
- Flexible modular system for a wide range of sizes up to 500 mL
- Low-well design is perfect for multi-neck flasks
Specifica
Specifica
| Materiale | Alluminio anodizzato |
| Tipo | Block |
| Include | base SHB250BASE |
| Dimensioni provetta | Medium |