Learn More
Sampling Systems™ Sub Sampler Pro
Codice prodotto. 15809361 Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Materiale:
Nickel-plated Brass
Stainless Steel
Dimensione della confezione:
Pezzo
Codice prodotto. 15809361
Marca: Sampling Systems™ 5072B500
Can sample from a set distance from the bottom of a container or alternatively at a mid point in a container or well. Sampling Systems™ Sub Sampler Pro is sometimes called a Bomb Sampler and it is used throughout the world for both chemical and environmental sampling.
  • Lower Sub Sampler Pro until the plunger assembly reaches the bottom of the container
  • When the plunger assembly hits the bottom, it is pushed up allowing the sample to enter the device
  • When the sampler is raised the plunger will drop down and seal the sampler
  • Lower the Sub Sampler Pro into the liquid
  • At the required depth pull on the control line to raise the plunger
  • This will allow the liquid to enter the sampler
  • Release the control Line and the plunger will drop down and seal the sample
Specifica
Product Type Sampler
Materiale Stainless Steel
Capacità (sistema metrico) 500 mL