Learn More
Sampling Systems™ Sub Sampler Pro
Sfoglia Tutto Sampling Systems Prodotti
Click to view available options
Materiale:
Nickel-plated Brass
Stainless Steel
Dimensione della confezione:
Pezzo
Beschreibung
- Lower Sub Sampler Pro until the plunger assembly reaches the bottom of the container
- When the plunger assembly hits the bottom, it is pushed up allowing the sample to enter the device
- When the sampler is raised the plunger will drop down and seal the sampler
- Lower the Sub Sampler Pro into the liquid
- At the required depth pull on the control line to raise the plunger
- This will allow the liquid to enter the sampler
- Release the control Line and the plunger will drop down and seal the sample
Spezifikation
Spezifikation
| Product Type | Sampler |
| Materiale | Nickel-plated Brass |
| Capacità (sistema metrico) | 500 mL |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur