Learn More
Metro™ Super Erecta™ Chrome Wire Shelf, Brite Finish
Codice prodotto. p-4247129
Click to view available options
Dimensioni (L x P):
24 x 14 in. (61 x 35.5 cm)
24 x 21 in. (61 x 53 cm)
30 x 18 in. (76 x 45.7 cm)
30 x 21 in. (76 x 53 cm)
36 x 14 in. (91.4 x 35.5 cm)
36 x 18 in. (91.4 x 45.7 cm)
36 x 21 in. (91.4 x 53 cm)
42 x 14 in. (106.6 x 35.5 cm)
42 x 21 in. (106.6 x 53 cm)
42 x 24 in. (106.6 x 61 cm)
48 x 21 in. (123 x 53 cm)
54 x 18 in. (137 x 45.7 cm)
54 x 21 in. (137 x 53 cm)
54 x 24 in. (137 x 61 cm)
60 x 18 in. (152.4 x 45.7 cm)
60 x 21 in. (152.4 x 53 cm)
72 x 14 in. (183 x 35.5 cm)
72 x 18 in. (183 x 45.7 cm)
72 x 21 in. (183 x 53 cm)
Capacità supporto carico (sistema imperiale):
600 lb.
800 lb.
Dimensione della confezione:
Pezzo
Codice prodotto. 15906621
Marca: Metro™ 2130BR
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Includes
Plastic sleeves and wedges.
Chrome wire shelf in brite finish is easily adjustable, offering multiple storage configurations when used with Metro Super Erecta shelving systems.
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Economical chromate finish for dry, low humidity environment
  • Lacquer coated finish provides the look of chrome
Specifica
Product Type Wire Shelf
Dimensioni (L x P) 30 x 21 in. (76 x 53 cm)
Finitura Brite
Da utilizzare con (apparecchiatura) Metro™ Super Erecta™ shelving system
Include Plastic sleeves and wedges
Lunghezza (sistema imperiale) 30 in.
Lunghezza (sistema metrico) 76 cm
Capacità supporto carico (sistema imperiale) 800 lb.
Capacità portata carico (sistema metrico) 363 kg
Larghezza (sistema imperiale) 21 in.
Larghezza (sistema metrico) 53 cm
Materiale Chrome