Learn More
OHAUS™ Support Stand
Codice prodotto. 15712389 Sfoglia Tutto Ohaus Prodotti
Click to view available options
Diametro (sistema metrico) asta:
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
Altezza (sistema metrico):
267 mm
305 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15712389
Marca: OHAUS™ 30392337
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
OHAUS™ Support Stand provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifica
Materiale Stainless Steel
Altezza (sistema metrico) 305 mm
Lunghezza (sistema imperiale) 12 in.
Lunghezza (sistema metrico) 305 mm
Lunghezza (sistema metrico) asta 914 mm
Larghezza (sistema imperiale) 16.50 in.
Larghezza (sistema metrico) 419 mm
Diametro (sistema metrico) asta 19 mm
Product Type Support Stand with Rod