Learn More
OHAUS™ Support Stand
Shop All Ohaus Products
Click to view available options
Diametro (sistema metrico) asta:
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
Altezza (sistema metrico):
267 mm
305 mm
Unit Size:
Pezzo
Description
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Specifications
| Materiale | Stainless Steel |
| Altezza (sistema metrico) | 267 mm |
| Lunghezza (sistema imperiale) | 10.50 in. |
| Lunghezza (sistema metrico) | 267 mm |
| Lunghezza (sistema metrico) asta | 711 mm |
| Larghezza (sistema imperiale) | 17.75 in. |
| Larghezza (sistema metrico) | 451 mm |
| Diametro (sistema metrico) asta | 16 mm |
| Product Type | Support Stand with Rod |