Learn More
OHAUS™ Support Stand Rod
Codice prodotto. 15702459 Sfoglia Tutto Ohaus Prodotti
Click to view available options
Lunghezza (sistema metrico):
460 mm
580 mm
710 mm
910 mm
1020 mm
1220 mm
1520 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15702459
Marca: OHAUS™ 30400038
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
OHAUS™ Support Stand Rod provides secure support for holding glassware at elevated positions.
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
TRUSTED_SUSTAINABILITY
Specifica
Tipo Rod
Materiale Stainless Steel
Descrizione Clamp, Support, Rod 71 cm, CLR-SPRODS071
Lunghezza (sistema metrico) 710 mm