Learn More
Testo™ Surface Probe
Click to view available options
Temperature Range (Metric):
0°C to 40°C
Dimensione della confezione:
Pezzo
Specifica
Specifica
| Min. Temperature (Metric) | 0°C |
| Temperature Range (Metric) | 0°C to 40°C |
| Accuracy | ±1°C |