missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Survivin Antibody (BIRC5/8987R), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-23789-20ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Survivin Monoclonal antibody specifically detects Survivin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifica
| Survivin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha | |
| Recombinant fragment (around aa1-100) of human Survivin protein (exact sequence is proprietary) | |
| 20 μg | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Astrocyte Markers, Cancer, Growth and Development, Innate Immunity, Mitotic Regulators, Phospho Specific, Stem Cells | |
| 332 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| BIRC5/8987R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto