Learn More
Syngene™ Gene Directory Track Database software
Sfoglia Tutto Syngene Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
Syngene Gbox
Dimensione della confezione:
Pezzo
Descrizione
- Used in conjunction with the GeneTools gel analysis package.
- Track classification with user defined fields.
- Genotyping capabilities.
- Powerful search facility for rapid identification of unknown tracks.
- Production of cluster analysis diagrams based on user defined parameters.
- VNTR table instantly calculated for any data set.
- Similarity matrix.
- Band matching to single or composite tracks.
- Dominance/Codominance marker identification with interface to JoinMap.
- GeneDirectory results create cluster analysis diagrams.
- Band match across multiple gels.
- Set up libraries with classification data.
- Make comparative quantifications.
- Share data between collaborating laboratories.
- Full report generation facility
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Syngene Gbox |
| Product Type | Software Track Database |