missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntenin 1 Antibody (OTI2H6) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-74418
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Syntenin 1 Monoclonal specifically detects Syntenin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifica
| Syntenin 1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-2000, Immunohistochemistry 1:150 | |
| MDA9, MDA-9, melanoma differentiation associated protein-9, Melanoma differentiation-associated protein 9, Pro-TGF-alpha cytoplasmic domain-interacting protein 18, Scaffold protein Pbp1, ST1, SYCLTACIP18, syndecan binding protein (syntenin), Syndecan-binding protein 1, syntenin-1 | |
| Mouse | |
| 32.3 kDa | |
| 100 μg | |
| 6386 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot, Immunohistochemistry | |
| OTI2H6 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| SDCBP | |
| Full length human recombinant protein of human SDCBP(NP_005616) produced in HEK293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto